Methyl-4-iodo-3-nitrobenzoate 98%

Supplier: Ambeed
Warning

Synonyms: 4-Iodobenzoic acid methyl ester, Methyl 4-iodo-3-nitrobenzoate

A264800-100G A264800-10G A264800-250MG A264800-1G A264800-25G A264800-5G
103812-690EA 7 USD
103812-690 103852-584 76977-046 76977-048 76977-050 77092-248 77092-250 77092-252
Methyl-4-iodo-3-nitrobenzoate 98%
Methyl-4-iodo-3-nitrobenzoate

Precaución: for Research Use Only

Formula: C₈H₆INO₄
MW: 307.04 g/mol
Melting Pt: 103…105 °C
Flash Pt: 171.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08064223
CAS Number: 89976-27-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR