Methyl-4-iodo-3-methoxybenzoate 98%

Supplier: Ambeed
Warning

A459179-100MG A459179-250MG A459179-25G A459179-5G A459179-1G
103827-894EA 25 USD
103827-894 103838-970 76725-688 76725-690 76842-838 76842-840 77307-420
Methyl-4-iodo-3-methoxybenzoate 98%
Methyl 4-iodo-3-methoxybenzoate

Precaución: for Research Use Only

Formula: C₉H₉IO₃
Boiling Pt: 324.3 °C (760 mm Hg)
Melting Pt: 48…51 °C
Storage Temperature: Ambient
MDL Number: MFCD06203960
CAS Number: 35387-92-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR