Methyl-4-hydroxyquinoline-2-carboxylate 95%
Supplier: Ambeed
|
Warning
|
A170724-100MG
A170724-10G
A170724-1G
A170724-5G
A170724-250MG
103806-532EA
18
USD
103806-532
103844-806
76998-190
76998-192
77081-920
77301-068
77881-316
Methyl-4-hydroxyquinoline-2-carboxylate 95%
Methyl 4-hydroxyquinoline-2-carboxylate
Precaución: for Research Use Only
Formula:
C₁₁H₉NO₃ Boiling Pt: 331.1 °C (760 mm Hg) Storage Temperature: Ambient |
MDL Number:
MFCD08693316 CAS Number: 5965-59-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...