Methyl-4-hydroxypyrimidine-5-carboxylate 96%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-hydroxypyrimidine-5-carboxylate

A115580-250MG A115580-25G A115580-1G
103798-588EA 9 USD
103798-588 103824-446 77094-214 77094-216 77311-262
Methyl-4-hydroxypyrimidine-5-carboxylate 96%
Methyl-4-hydroxypyrimidine-5-carboxylate

Precaución: for Research Use Only

Formula: C₆H₆N₂O₃
MW: 154.1200 g/mol
Boiling Pt: 256.9 °C (760 mm Hg)
Melting Pt: 209…210 °C
Storage Temperature: Ambient
MDL Number: MFCD01632117
CAS Number: 4774-35-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR