Methyl-4-hydroxycyclohexanecarboxylate 96%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 4-hydroxycyclohexanecarboxylate
A195525-10G
A195525-1G
A195525-5G
A195525-25G
103816-764EA
9
USD
103816-764
103847-376
76721-274
77085-332
77085-334
77302-474
Methyl-4-hydroxycyclohexanecarboxylate 96%
Methyl-4-hydroxycyclohexanecarboxylate
Precaución: for Research Use Only
Formula:
C₈H₁₄O₃ MW: 158.2 g/mol Boiling Pt: 233.2 °C (760 mm Hg) Flash Pt: 94 °C Storage Temperature: Ambient |
MDL Number:
MFCD00729087 CAS Number: 17449-76-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...