Methyl 4-hydroxy-7-phenoxyisoquinoline-3-carboxylate 97%

Supplier: Ambeed
Warning

A110786-250MG A110786-25G A110786-1G A110786-5G
103797-732EA 31 USD
103797-732 103823-540 77233-536 77233-538 77284-042 77284-044
Methyl 4-hydroxy-7-phenoxyisoquinoline-3-carboxylate 97%
Methyl 4-hydroxy-7-phenoxyisoquinoline-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₇H₁₃NO₄
MW: 295.29 g/mol
Boiling Pt: 534.7±45.0 °C (760 mm Hg)
Flash Pt: 277.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD29075436
CAS Number: 1455091-10-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR