4-Hydroxy-5-methoxy-2-nitrobenzoic acid methylester 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-hydroxy-5-methoxy-2-nitrobenzoate

A101755-250MG A101755-1G A101755-5G A101755-25G
103796-118EA 64 USD
103796-118 103821-806 77160-214 77308-712 77308-714 77682-937
4-Hydroxy-5-methoxy-2-nitrobenzoic acid methylester 97%
4-Hydroxy-5-methoxy-2-nitrobenzoic acid methylester

Precaución: For research use only.

Formula: C₉H₉NO₆
MW: 227.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09833001
CAS Number: 27883-60-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR