Methyl-4-hydroxy-3-methylbenzoate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-hydroxy-3-methylbenzoate

A148562-10G A148562-1G A148562-25G A148562-5G
103803-942EA 21 USD
103803-942 103842-058 77286-046 77299-666 77299-668 77299-670
Methyl-4-hydroxy-3-methylbenzoate 97%
Methyl-4-hydroxy-3-methylbenzoate

Precaución: for Research Use Only

Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 282.9 °C (760 mm Hg)
Melting Pt: 124…125 °C
Flash Pt: 122.2 °C
Storage Temperature: Ambient
MDL Number: MFCD06203848
CAS Number: 42113-13-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR