Methyl 4-hydrazinylbenzoate hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A462348-1G
A462348-25G
A462348-5G
A462348-250MG
103827-936EA
14
USD
103827-936
103839-012
76842-896
76842-898
76842-900
77291-800
Methyl 4-hydrazinylbenzoate hydrochloride 97%
Methyl 4-hydrazinylbenzoate hydrochloride
Precaución: for Research Use Only
Formula:
C₈H₁₁ClN₂O₂ Boiling Pt: 923.4 °C (760 mm Hg) Flash Pt: 512.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD01325154 CAS Number: 6296-89-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...