Methyl-4H-thieno[3,2-b]pyrrole-5-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 4H-thieno[3,2-b]pyrrole-5-carboxylate

A128471-100G A128471-100MG A128471-10G A128471-250MG A128471-1G A128471-5G
103801-134EA 39 USD
103801-134 77026-936 77026-938 77026-940 77026-942 77313-454 77313-456
Methyl-4H-thieno[3,2-b]pyrrole-5-carboxylate 98%
Methyl-4H-thieno[3,2-b]pyrrole-5-carboxylate

Precaución: for Research Use Only

Formula: C₈H₇NO₂S
MW: 181.2117 g/mol
Melting Pt: 143…145 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06797484
CAS Number: 82782-85-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR