Methyl 4-formylbicyclo[2.2.2]octane-1-carboxylate ≥95%

Supplier: Ambeed
Warning

Synonyms: Methyl4-Formylbicyclo[2.2.2]octane-1-carboxylate

A146764-100MG A146764-250MG
103803-698EA 379 USD
103803-698 103841-794 77611-354 77683-253
Methyl 4-formylbicyclo[2.2.2]octane-1-carboxylate ≥95%
Methyl 4-formylbicyclo[2.2.2]octane-1-carboxylate

Precaución: For research use only.

Formula: C₁₁H₁₆O₃
MW: 196.24 g/mol
Storage Temperature: Freezer
MDL Number: MFCD19105426
CAS Number: 94994-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR