Methyl-4-formylbenzoate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl terephthalaldehydate, Methyl 4-formylbenzoate

A563203-1000G A563203-25G A563203-100G A563203-10G A563203-1G A563203-500G
103810-456EA 69 USD
103810-456 76843-996 76843-998 77266-364 77266-366 77266-368 77266-370
Methyl-4-formylbenzoate 98%
Methyl-4-formylbenzoate

Precaución: for Research Use Only

Formula: C₉H₈O₃
MW: 164.16 g/mol
Boiling Pt: 265 °C (760 mm Hg)
Melting Pt: 59…63 °C
Flash Pt: 128 °C
Storage Temperature: Ambient
MDL Number: MFCD00006950
CAS Number: 1571-08-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR