Methyl 4-fluorothiophene-2-carboxylate 95%

Supplier: Ambeed
Warning

A324231-100MG A324231-1G A324231-5G A324231-250MG
103825-366EA 231 USD
103825-366 103836-308 76817-082 76839-228 76839-230 77883-878
Methyl 4-fluorothiophene-2-carboxylate 95%
Methyl 4-fluorothiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₆H₅FO₂S
Storage Temperature: Refrigerator
MDL Number: MFCD11618006
CAS Number: 32431-75-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR