Methyl-4-fluorobenzoate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-fluorobenzoate, 4-Fluorobenzoic acid emthyl ester

A352611-1000G A352611-100G A352611-500G A352611-25G A352611-5G
103810-204EA 112 USD
103810-204 103837-072 76840-182 77240-022 77240-024 77240-026 77306-634 77306-636
Methyl-4-fluorobenzoate 98%
Methyl-4-fluorobenzoate

Precaución: for Research Use Only

Formula: C₈H₇FO₂
MW: 154.14 g/mol
Boiling Pt: 190.4 °C (760 mm Hg)
Flash Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD00017959
CAS Number: 403-33-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR