Methyl-4-fluoro-2-methoxybenzoate 97%

Supplier: Ambeed
Warning

A115180-10G A115180-250MG A115180-1G
103798-526EA 13 USD
103798-526 103824-372 77284-462 77284-464 77311-186
Methyl-4-fluoro-2-methoxybenzoate 97%
Methyl-4-fluoro-2-methoxybenzoate

Precaución: for Research Use Only

Formula: C₉H₉FO₃
Boiling Pt: 231.646 °C (760 mm Hg)
Melting Pt: 134…136 °C at N/A mmHg
Flash Pt: 91.331 °C
Storage Temperature: Ambient
MDL Number: MFCD06203990
CAS Number: 204707-42-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR