Methyl-4-fluorosalicylate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-fluoro-2-hydroxybenzoate

A380284-10G A380284-250MG A380284-1G
103826-666EA 9 USD
103826-666 103837-702 76724-864 76724-866 76841-024
Methyl-4-fluorosalicylate 98%
Methyl 4-fluorosalicylate

Precaución: for Research Use Only

Formula: C₈H₇FO₃
Boiling Pt: 224.7 °C (760 mm Hg)
Melting Pt: 38…40 °C
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD02093965
CAS Number: 392-04-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR