Methyl 4-fluoro-1H-indazole-6-carboxylate 95%
Supplier: Ambeed
|
Warning
|
A155214-100MG
A155214-10G
A155214-250MG
A155214-5G
103804-732EA
99
USD
103804-732
103842-926
76984-794
76984-796
76984-798
77318-650
Methyl 4-fluoro-1H-indazole-6-carboxylate 95%
Methyl 4-fluoro-1H-indazole-6-carboxylate
Precaución: for Research Use Only
Formula:
C₉H₇FN₂O₂ MW: 194.16 g/mol Boiling Pt: 357.6±27.0 °C (760 mm Hg) Flash Pt: 170.1 °C Storage Temperature: Ambient |
MDL Number:
MFCD08272209 CAS Number: 885521-44-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...