Methyl 4-cyano-3-fluorobenzoate 97%

Supplier: Ambeed
Warning

A318223-1G A318223-25G A318223-5G A318223-250MG
103825-164EA 15 USD
103825-164 103836-096 76838-948 76838-950 76838-952 77099-798
Methyl 4-cyano-3-fluorobenzoate 97%
Methyl 4-cyano-3-fluorobenzoate

Precaución: for Research Use Only

Formula: C₉H₆FNO₂
Boiling Pt: 294.9 °C (760 mm Hg)
Flash Pt: 132.1 °C
Storage Temperature: Ambient
MDL Number: MFCD09264509
CAS Number: 268734-34-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR