Methyl 4-cyano-2-methylbenzoate 97%

Supplier: Ambeed
Warning

A186854-100MG A186854-10G A186854-250MG A186854-1G A186854-25G A186854-5G
103815-970EA 15 USD
103815-970 103846-556 76974-686 76974-688 76974-690 77084-244 77084-246 77084-248
Methyl 4-cyano-2-methylbenzoate 97%
Methyl 4-cyano-2-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₉NO₂
MW: 175.184 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18399110
CAS Number: 103261-67-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR