Methyl 4-cyano-2-fluoro-5-nitrobenzoate 95%

Supplier: Ambeed
Warning

A382895-1G A382895-100MG A382895-250MG
103826-722EA 135 USD
103826-722 76841-080 77168-088 77168-090
Methyl 4-cyano-2-fluoro-5-nitrobenzoate 95%
Methyl 4-cyano-2-fluoro-5-nitrobenzoate

Precaución: for Research Use Only

Formula: C₉H₅FN₂O₄
Storage Temperature: Ambient
MDL Number: MFCD26390065
CAS Number: 1149388-51-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR