Methyl-4-(chlorosulfonyl)benzoate 93%

Supplier: Ambeed
Danger

Synonyms: Methyl 4-chlorosulphonylbenzoate

A556355-10G A556355-250MG A556355-1G A556355-5G
103828-720EA 19 USD
103828-720 103839-856 77292-476 77292-478 77307-798 77307-800
Methyl-4-(chlorosulfonyl)benzoate 93%
Methyl 4-(chlorosulphonyl)benzoate

Precaución: for Research Use Only

Formula: C₈H₇ClO₄S
MW: 234.6568 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00627554
CAS Number: 69812-51-7
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR