Methyl 4-chloroquinoline-7-carboxylate 98%

Supplier: Ambeed
Warning

A119275-250MG A119275-1G
103799-190EA 68 USD
103799-190 76810-364 77311-862
Methyl 4-chloroquinoline-7-carboxylate 98%
Methyl 4-chloroquinoline-7-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₈ClNO₂
MW: 221.64 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18381419
CAS Number: 178984-69-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR