Methyl 4-chloro-5-fluoroanthranilate 97%

Supplier: Ambeed
Warning

A760019-5G A760019-100MG A760019-250MG
103840-880EA 137 USD
103840-880 76845-112 76993-110 76993-112
Methyl 4-chloro-5-fluoroanthranilate 97%
Methyl 4-chloro-5-fluoroanthranilate

Precaución: for Research Use Only

Formula: C₈H₇ClFNO₂
MW: 203.598 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15523631
CAS Number: 104901-79-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR