Methyl-4-chloro-3-nitrobenzoate 98%

Supplier: Ambeed
Warning

Synonyms: 4-Chloro-3-nitrobenzoic acid methyl ester, Methyl 4-chloro-3-nitrobenzate

A260549-10G A260549-1G A260549-5G A260549-100G
103852-382EA 6 USD
103852-382 77000-920 77000-922 77091-966 77672-324
Methyl-4-chloro-3-nitrobenzoate 98%
Methyl-4-chloro-3-nitrobenzoate

Precaución: for Research Use Only

Formula: C₈H₆ClNO₄
Boiling Pt: 313.2 °C (760 mm Hg)
Melting Pt: 80 °C
Flash Pt: 143.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00016989
CAS Number: 14719-83-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR