Methyl 4-chloro-3-methylpicolinate 97%

Supplier: Ambeed
Warning

A127883-1G A127883-5G
103801-032EA 82 USD
103801-032 77298-248 77298-250
Methyl 4-chloro-3-methylpicolinate 97%
Methyl 4-chloro-3-methylpicolinate

Precaución: for Research Use Only

Formula: C₈H₈ClNO₂
MW: 185.61 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09839160
CAS Number: 1260764-76-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR