Methyl-4-chloro-3-methoxy-5-nitrobenzenecarboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-chloro-3-methoxy-5-nitrobenzoate

A236501-10G A236501-250MG A236501-500G A236501-1G A236501-5G A236501-100G
103811-102EA 10 USD
103811-102 103850-874 76976-350 76976-352 76976-354 76976-356 77089-922 77089-924 77672-104
Methyl-4-chloro-3-methoxy-5-nitrobenzenecarboxylate 97%
Methyl-4-chloro-3-methoxy-5-nitrobenzenecarboxylate

Precaución: for Research Use Only

Formula: C₉H₈ClNO₅
Boiling Pt: 366.2±37.0 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD16140357
CAS Number: 63603-09-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR