Methyl 4-chloro-2-cyanobenzoate 95%

Supplier: Ambeed
Warning

A179890-100MG A179890-1G
103807-486EA 103 USD
103807-486 103845-840 76720-840 77083-316
Methyl 4-chloro-2-cyanobenzoate 95%
Methyl 4-chloro-2-cyanobenzoate

Precaución: for Research Use Only

Formula: C₉H₆ClNO₂
MW: 195.6 g/mol
Boiling Pt: 305.8 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD08234732
CAS Number: 58331-97-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR