Methyl 4-chloro-2,6-difluorobenzoate 95%

Supplier: Ambeed
Warning

A816471-100MG A816471-1G A816471-250MG A816471-5G
103841-108EA 297 USD
103841-108 77019-494 77019-496 77019-498 77308-372
Methyl 4-chloro-2,6-difluorobenzoate 95%
Methyl 4-chloro-2,6-difluorobenzoate

Precaución: for Research Use Only

Formula: C₈H₅ClF₂O₂
MW: 206.57 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06204170
CAS Number: 773134-12-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR