Methyl-4-chloro-1H-pyrrolo[2,3-b]pyridine-2-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-chloro-1H-pyrrolo[2,3-b]pyridine-2-carboxylate

A336454-100MG A336454-10G A336454-250MG A336454-1G A336454-5G
103825-720EA 27 USD
103825-720 103836-694 76788-246 76788-248 76788-250 76839-702 76839-704
Methyl-4-chloro-1H-pyrrolo[2,3-b]pyridine-2-carboxylate 95%
Methyl-4-chloro-1H-pyrrolo[2,3-b]pyridine-2-carboxylate

Precaución: for Research Use Only

Formula: C₉H₇ClN₂O₂
MW: 210.6200 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09839319
CAS Number: 871583-23-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR