Methyl 4-chloro-1H-indole-2-carboxylate 97%
Supplier: Ambeed
|
Warning
|
A107191-100MG
A107191-250MG
A107191-1G
103797-150EA
51
USD
103797-150
103822-912
77008-684
77008-686
77309-768
Methyl 4-chloro-1H-indole-2-carboxylate 97%
Methyl 4-chloro-1H-indole-2-carboxylate
Precaución: for Research Use Only
Formula:
C₁₀H₈ClNO₂ Boiling Pt: 365.5 °C (760 mm Hg) Melting Pt: 146…147 °C Flash Pt: 174.8 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD04966957 CAS Number: 230291-43-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...