Methyl 4-butyramido-3-methylbenzoate 97%

Supplier: Ambeed
Warning

A271665-250MG A271665-1G A271665-5G
103813-056EA 26 USD
103813-056 103852-968 77289-456 77305-088 77305-090
Methyl 4-butyramido-3-methylbenzoate 97%
Methyl 4-butyramido-3-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₃H₁₇NO₃
Boiling Pt: 397.5 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD09839029
CAS Number: 301533-59-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR