Methyl 4-bromothiophene-2-carboxylate 98%
Supplier: Ambeed
|
Warning
|
A258051-100G
A258051-10G
A258051-1G
A258051-25G
A258051-5G
103812-356EA
7
USD
103812-356
103852-228
76722-720
76722-722
77091-752
77091-754
77091-756
Methyl 4-bromothiophene-2-carboxylate 98%
Methyl 4-bromothiophene-2-carboxylate
Precaución: for Research Use Only
Formula:
C₆H₅BrO₂S Boiling Pt: 255 °C (760 mm Hg) Flash Pt: 108 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD06203668 CAS Number: 62224-16-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...