Methyl-4-bromofuran-2-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-bromofuran-2-carboxylate

A161256-100G A161256-250MG A161256-1G A161256-25G A161256-5G A161256-10G
103805-472EA 13 USD
103805-472 103843-710 76720-138 76720-140 77319-674 77319-676 77319-678 77600-286
Methyl-4-bromofuran-2-carboxylate 98%
Methyl-4-bromofuran-2-carboxylate

Precaución: for Research Use Only

Formula: C₆H₅BrO₃
MW: 205.0100 g/mol
Boiling Pt: 228 °C (760 mm Hg)
Flash Pt: 92 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00092559
CAS Number: 58235-80-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR