Methyl-4-bromobutyrate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-bromobutanoate

A360958-1000G A360958-100G A360958-10G A360958-1G A360958-500G A360958-25G A360958-5G
103837-288EA 6 USD
103837-288 76724-592 76724-594 76724-596 76724-598 76724-600 76840-506 77306-720
Methyl-4-bromobutyrate 98%
Methyl-4-bromobutyrate

Precaución: for Research Use Only

Formula: C₅H₉BrO₂
Boiling Pt: 186.5 °C (760 mm Hg)
Flash Pt: 186 °C
Storage Temperature: Ambient
MDL Number: MFCD00041482
CAS Number: 4897-84-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR