Methyl-4-bromobenzo[b]thiophene-2-carboxylate 97%

Supplier: Ambeed
Warning

A198966-10G A198966-250MG A198966-5G
103817-060EA 30 USD
103817-060 103847-686 76986-370 76986-372 77085-712
Methyl-4-bromobenzo[b]thiophene-2-carboxylate 97%
Methyl 4-bromobenzo[b]thiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₇BrO₂S
Melting Pt: 105…108 °C
Storage Temperature: Ambient
MDL Number: MFCD07371547
CAS Number: 360575-29-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR