Methyl-4-bromo-5-methylisoxazole-3-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-bromo-5-methylisoxazole-3-carboxylate

A240901-100MG A240901-250MG A240901-1G
103811-376EA 216 USD
103811-376 103851-182 77000-430 77000-432 77304-206
Methyl-4-bromo-5-methylisoxazole-3-carboxylate 95%
Methyl-4-bromo-5-methylisoxazole-3-carboxylate

Precaución: for Research Use Only

Formula: C₆H₆BrNO₃
MW: 220.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12028409
CAS Number: 850832-54-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR