Methyl-4-bromo-3-nitrobenzoate 97%

Supplier: Ambeed
Warning

Synonyms: 4-Bromobenzoic acid methyl ester, Methyl 4-bromo-3-nitrobenzoate

A101448-100G A101448-10G A101448-25G A101448-1G A101448-5G
103795-978EA 6 USD
103795-978 103821-662 76765-170 76765-172 76765-174 77282-738 77297-104
Methyl-4-bromo-3-nitrobenzoate 97%
Methyl-4-bromo-3-nitrobenzoate

Precaución: for Research Use Only

Formula: C₈H₆BrNO₄
Boiling Pt: 320.9 °C (760 mm Hg)
Melting Pt: 102…103 °C
Flash Pt: 147.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00553089
CAS Number: 2363-16-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR