Methyl-4-bromo-3-methylbenzoate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-bromo-3-methylbenzoate

A206269-100G A206269-10G A206269-500G A206269-25G A206269-5G A206269-1G
103817-698EA 9 USD
103817-698 103848-368 76999-500 76999-502 76999-504 77086-614 77086-616 77302-984
Methyl-4-bromo-3-methylbenzoate 98%
Methyl 4-bromo-3-methylbenzoate

Precaución: for Research Use Only

Formula: C₉H₉BrO₂
MW: 229.07 g/mol
Melting Pt: 38…44 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00673014
CAS Number: 148547-19-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR