Methyl-4-bromo-3-methoxybenzoate 97%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-3-methoxybenzoic acid methyl ester, Methyl 4-bromo-3-methoxybenzoate

A108480-100G A108480-10G A108480-250MG A108480-25G A108480-1G A108480-5G
103797-356EA 6 USD
103797-356 103823-136 76717-960 76717-962 76717-964 76717-966 77309-978 77309-980
Methyl-4-bromo-3-methoxybenzoate 97%
Methyl-4-bromo-3-methoxybenzoate

Precaución: for Research Use Only

Formula: C₉H₉BrO₃
MW: 245.07 g/mol
Boiling Pt: 306 °C (760 mm Hg)
Melting Pt: 58…61 °C
Flash Pt: 138.9 °C
Storage Temperature: Ambient
MDL Number: MFCD08436018
CAS Number: 17100-63-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR