Methyl 4-bromo-3-hydroxy-2-methylbenzoate 97%

Supplier: Ambeed
Warning

A224158-1G A224158-5G A224158-100MG A224158-250MG
103819-090EA 36 USD
103819-090 103849-980 77088-618 77088-620 77139-896 77139-898
Methyl 4-bromo-3-hydroxy-2-methylbenzoate 97%
Methyl 4-bromo-3-hydroxy-2-methylbenzoate

Precaución: for Research Use Only

Formula: C₉H₉BrO₃
Boiling Pt: 318.2±37.0 °C (760 mm Hg)
Flash Pt: 146.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD26389396
CAS Number: 1149388-19-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR