Methyl 4-bromo-3-fluorothiophene-2-carboxylate 97%

Supplier: Ambeed
Warning

A276628-100MG A276628-250MG A276628-1G
103813-312EA 583 USD
103813-312 103853-248 77276-878 77276-880 77305-240
Methyl 4-bromo-3-fluorothiophene-2-carboxylate 97%
Methyl 4-bromo-3-fluorothiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₆H₄BrFO₂S
Boiling Pt: 267.1 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD10566744
CAS Number: 395664-56-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR