Methyl-4-bromo-3-fluorobenzoate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-bromo-3-fluorobenzoate

A226722-100G A226722-10G A226722-250MG A226722-1G A226722-25G A226722-5G
103819-264EA 6 USD
103819-264 103850-168 76874-152 76874-154 76874-156 77088-912 77088-914 77088-916
Methyl-4-bromo-3-fluorobenzoate 98%
Methyl-4-bromo-3-fluorobenzoate

Precaución: for Research Use Only

Formula: C₈H₆BrFO₂
Boiling Pt: 270.1 °C (760 mm Hg)
Melting Pt: 64…66 °C
Flash Pt: 117.14 °C
Storage Temperature: Ambient
MDL Number: MFCD08436044
CAS Number: 849758-12-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR