Methyl 4-Bromo-3-(bromomethyl)benzoate 95%

Supplier: Ambeed
Danger

A235223-1G A235223-5G A235223-100MG A235223-250MG
103810-996EA 103 USD
103810-996 103850-760 77089-762 77089-764 77165-662 77165-664
Methyl 4-Bromo-3-(bromomethyl)benzoate 95%
Methyl 4-Bromo-3-(bromomethyl)benzoate

Precaución: for Research Use Only

Formula: C₉H₈Br₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD11858300
CAS Number: 142031-67-2
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR