Methyl-4-bromo-2-methylbenzoate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-bromo-2-methylbenzoate

A487663-100G A487663-10G A487663-500G A487663-25G A487663-5G A487663-1G
103828-188EA 9 USD
103828-188 103839-262 76789-752 76789-754 76789-756 76843-210 76843-212 77307-576
Methyl-4-bromo-2-methylbenzoate 98%
Methyl 4-bromo-2-methylbenzoate

Precaución: for Research Use Only

Formula: C₉H₉BrO₂
MW: 229.0706 g/mol
Boiling Pt: 273.6 °C (760 mm Hg)
Flash Pt: 119.286 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09954957
CAS Number: 99548-55-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR