Methyl-4-bromo-2-methoxybenzoate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-bromo-2-methoxybenzoate, 4-Bromo-2-methoxybenzoic acid methyl ester

A145284-100G A145284-10G A145284-1G A145284-25G A145284-5G
103803-516EA 6 USD
103803-516 103841-598 76997-024 76997-026 76997-028 77316-878 77316-880 77316-882
Methyl-4-bromo-2-methoxybenzoate 98%
Methyl-4-bromo-2-methoxybenzoate

Precaución: for Research Use Only

Formula: C₉H₉BrO₃
MW: 245.07
Boiling Pt: 100…108 °C (760 mm Hg)
Melting Pt: 33 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD06797867
CAS Number: 139102-34-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR