Methyl-4-bromosalicylate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-bromo-2-hydroxybenzoate

A103047-10G A103047-250MG A103047-25G A103047-1G A103047-5G
103796-474EA 6 USD
103796-474 103822-176 76994-622 76994-624 76994-626 77309-044 77309-046
Methyl-4-bromosalicylate 98%
Methyl 4-bromosalicylate

Precaución: for Research Use Only

Formula: C₈H₇BrO₃
MW: 231.0434 g/mol
Melting Pt: 41…42 °C
Flash Pt: 126 °C
Storage Temperature: Ambient
MDL Number: MFCD07780736
CAS Number: 22717-56-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR