Methyl-4-bromo-2-fluorobenzoate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-bromo-2-fluorobenzoate

A321711-25G A321711-100G A321711-10G A321711-500G A321711-5G
103836-222EA 10 USD
103836-222 76839-134 77002-872 77002-874 77002-876 77306-250
Methyl-4-bromo-2-fluorobenzoate 98%
Methyl-4-bromo-2-fluorobenzoate

Precaución: for Research Use Only

Formula: C₈H₆BrFO₂
MW: 233.04 g/mol
Melting Pt: 58…60 °C
Flash Pt: 118.7 °C
Storage Temperature: Ambient
MDL Number: MFCD06203731
CAS Number: 179232-29-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR