Methyl 4-bromo-2-chloro-6-methylbenzoate 98%

Supplier: Ambeed
Warning

A360616-250MG A360616-1G A360616-25G A360616-5G
103826-262EA 27 USD
103826-262 103837-272 76876-392 77306-712 77306-714 77306-716
Methyl 4-bromo-2-chloro-6-methylbenzoate 98%
Methyl 4-bromo-2-chloro-6-methylbenzoate

Precaución: for Research Use Only

Formula: C₉H₈BrClO₂
MW: 263.52 g/mol
Flash Pt: 140.167 °C
Storage Temperature: Ambient
MDL Number: MFCD09260942
CAS Number: 877149-10-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR