Methyl-4-bromo-2-(bromomethyl)benzoate 97%

Supplier: Ambeed
Danger

A103175-100G A103175-10G A103175-250MG A103175-25G A103175-500G A103175-1G A103175-5G
103796-498EA 14 USD
103796-498 103822-202 77008-450 77008-452 77008-454 77008-456 77008-458 77309-078 77309-080
Methyl-4-bromo-2-(bromomethyl)benzoate 97%
Methyl 4-bromo-2-(bromomethyl)benzoate

Precaución: for Research Use Only

Formula: C₉H₈Br₂O₂
Boiling Pt: 364.2 °C (760 mm Hg)
Flash Pt: 174.103 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04114319
CAS Number: 78471-43-9
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR