Methyl-4-bromo-2,6-difluorobenzoate 98%

Supplier: Ambeed
Warning

A323436-25G A323436-5G A323436-100G A323436-10G A323436-1G
103836-280EA 15 USD
103836-280 76839-196 76839-198 77114-740 77114-742 77114-744
Methyl-4-bromo-2,6-difluorobenzoate 98%
Methyl 4-bromo-2,6-difluorobenzoate

Precaución: for Research Use Only

Formula: C₈H₅BrF₂O₂
Boiling Pt: 276.1 °C (760 mm Hg)
Melting Pt: 41…43 °C
Flash Pt: 120.8 °C
Storage Temperature: Ambient
MDL Number: MFCD06204169
CAS Number: 773134-11-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR